missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITGA6X1A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20231-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
E3 ubiquitin-protein ligase that mediates the ubiquitination of p53/TP53 in the late response to DNA damage, and acts as a positive regulator of p53/TP53 stability, thereby regulating the G1/S DNA damage checkpoint. May act by catalyzing the formation of short polyubiquitin chains on p53/TP53 that are not targeted to the proteasome. In response to ionizing radiation, interacts with MDM2 and enhances p53/TP53 ubiquitination, possibly by restricting MDM2 from extending polyubiquitin chains on ubiquitinated p53/TP53. Plays a role in RPA-mediated DNA damage signaling and repair.Spezifikation
ITGA6X1A | |
Polyclonal | |
Unconjugated | |
ITGA6 | |
CD49f, Integrin alpha 6, integrin, alpha 6, ITGA6, ITGA6B, VLA 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16403, 3655 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P23229, Q61739 | |
Itga6 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |