Learn More
IPPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12603-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a kinase that phosphorylates position 2 of inositol-1, 3, 4, 5, 6-pentakisphosphate to form inositol-1, 2, 3, 4, 5, 6-hexakisphosphate (InsP6). InsP6 has a variety of functions, including stimulation of DNA repair, endocytosis, and mRNA export.Spezifikation
IPPK | |
Polyclonal | |
Unconjugated | |
IPPK | |
C9orf12, Ins(1,3,4,5,6)P5 2 kinase, InsP5 2 kinase, INSP5K2, IP5K, IPK1, IPK1 homolog, IPPK, KIAA0699 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
306808, 64768, 75678 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5PXE9, Q6P1C1, Q9H8X2 | |
IPPK | |
IPPK Fusion Protein Ag3278 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |