Learn More
Thermo Scientific™ Invitrogen™ EVOS™ FL Auto 2 Imaging System
Artikelnummer. 15736152
Click to view available options
Packungsgröße:
1 Stück
Artikelnummer. 15736152
Marke: Thermo Scientific™ AMAFD2000
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
Alternative Produkte anzeigen
Designed with advanced capabilities to simplify demanding cell-based imaging applications such as live-cell imaging, image tiling, and Z-stacking. Bring high performance right to your lab bench with the new Invitrogen™ EVOS™ FL Auto 2 Imaging System.
  • Speed: scan a 96-well plate in 3 fluorescent channels in less than 5 minutes
  • Flexibility: customize the system with more than 20 user-changeable LED light cubes, dual cameras (monochrome and color), a variety of objectives ranging from 1.25x to 100x, and multiple vessel holders
  • Time-lapse live-cell imaging: option for precise control of temperature, humidity, and gases for normoxic or hypoxic conditions allows a wide range of biological studies under physiological conditions with an onstage incubator
  • Area view: move rapidly and seamlessly between low-magnification, single-field mode and high-magnification scan mode to easily define and capture the area of interest
  • Automation: time-saving features such as autofocus, rapid stage movement, and automated routines help reduce time to complete experiments, allowing high throughput, high data quality, and improved experimental reproducibility
  • Data analysis: optional advanced software package for quantitative and statistical analysis
TRUSTED_SUSTAINABILITY
Spezifikation
Zur Verwendung mit (Anwendung) Bildgebung lebender Zellen
Zur Verwendung mit (Geräte) EVOS Light Cubes, EVOS Onstage Incubator