missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INTS8 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18802-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a component of prolyl 4-hydroxylase, a key enzyme in collagen synthesis composed of two identical alpha subunits and two beta subunits. The encoded protein is one of several different types of alpha subunits and provides the major part of the catalytic site of the active enzyme. In collagen and related proteins, prolyl 4-hydroxylase catalyzes the formation of 4-hydroxyproline that is essential to the proper three-dimensional folding of newly synthesized procollagen chains. Alternatively spliced transcript variants encoding different isoforms have been described.Spezifikation
INTS8 | |
Polyclonal | |
Unconjugated | |
INTS8 | |
C8orf52, INT8, integrator complex subunit 8, INTS8, Protein kaonashi 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55656 | |
Antibody | |
IgG |
Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q75QN2 | |
INTS8 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Liquid |