missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement (100X)

Artikelnummer. 30284786 Alle ansehen Gibco Produkte
Ansicht ändern
Click to view available options
Menge:
10 ml
100 ml
Packungsgröße:
10 Milliliter
100 Milliliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
30284786 10 ml 10 Milliliter
30284105 100 ml 100 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30284786 Lieferant Gibco™ Lieferanten-Nr. A4000046401
Neugier zahlt sich aus
Verwenden Sie den Aktionscode 27966
Alle Werbeaktionen anzeigen
 Eingeschränkter Artikel
Lager vorrätig
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement is an animal-origin-free (AOF) basal medium supplement that is used to reduce the amount of FBS needed to culture cells.

Gibco Insulin-Transferrin-Selenium-Ethanolamine (ITS-X) Select Supplement is an animal-origin-free (AOF) basal medium supplement. The insulin and transferrin in the ITS-X Select formulation are produced in non-mammalian recombinant expression systems.

ITS-X is used as a basal medium supplement to reduce the amount of fetal bovine serum (FBS) used to culture cells. It is routinely used, in combination with 2–4% FBS, to support cell types that require ethanolamine, such as bronchial epithelial cells.

Insulin promotes glucose and amino acid uptake, lipogenesis, intracellular transport, and the synthesis of proteins and nucleic acids. Transferrin is an iron carrier and may also help reduce toxic levels of oxygen radicals and peroxide. Selenium, as sodium selenite, is a co-factor for glutathione peroxidase and other proteins that is used as an antioxidant in media. Ethanolamine is a precursor of phospho-glycerides, which are essential to the structure of the plasma membrane and cellular organelles.

The product formulation contains the following components:

  • Insulin (1000.0 mg/L)
  • Transferrin (550.0 mg/L)
  • Sodium selenite (0.67 mg/L)
  • Ethanolamine (200.0 mg/L)

cGMP manufacturing and quality system

ITS-X Select Supplement is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.

TRUSTED_SUSTAINABILITY

Spezifikation

Konzentration 100 X
Kulturtyp Mammalian Cell Culture
Form Liquid
Produkttyp Cell Culture Supplement
Sterilität Sterile-filtered
Sterilization Method Sterile-filtered
Endotoxin-Gehalt ≤ 1.00 EU/mL
Fertigungsqualität, Herstellungsqualität cGMP-compliant under the ISO 13485 standard
Osmolalität 275 to 310 mOsm/kg
pH 7.8 to 9.5
Menge 10 ml
Versandbedingung Room Temperature
Quelle Animal Origin-free
Mehr anzeigen Weniger anzeigen

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.