missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InPro™ 4800 SG Pt1000 Analog Electrode

A top-of-the-line combined pH and temperature electrode designed to handle high-temperature and high-pressure dirty chemical applications.

Marke:  Mettler Toledo™ 52003542

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 12737661

  • 935.00 CHF / 1 Stück


Voraussichtliches Versanddatum: 17-06-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Solution ground (SG) feature improves grounding and effectively eliminates the risk of electrostatic interference, as well as improving advanced diagnostic functions
  • Strong resistance to oxidizing media, solvents, acid and alkali solutions makes it suitable for highly demanding industrial applications including chemical processing, chlor-alkali, pulp and paper, dyes and pigments and sugar processing
  • Very long diffusion path using two electrolyte chambers
  • PTFE annular junction expels dirt
  • Watertight connector (IP 68), integral temperature sensor
  • ATEX and FM certified for hazardous areas
  • High pressure and temperature rating of 12 barg at 130°C

Spezifikation

Spezifikation

Watertight IP 68, ISM: K8S cable
12 mm
120 mm
Reference system Ag/AgCl
Analog Electrode
Operating pressure of 12 barg at 130°C
Dirty chemical applications
Glass shaft and membrane
-5°C to +130°C
0 to 14
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation