missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ InPro™ 3100 Pt1000 Analog Electrode

A combined pH electrode and temperature sensor designed specially for in-line pH measurements in bio-processes where CIP and SIP are used.

Marke:  Mettler Toledo™ 52000666

Additional Details : Gewicht : 0.10000kg

Artikelnummer. 12717531

  • 1050.00 CHF / 1 Stück
Verfügbar ab: 18-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Rugged gel-filled electrode leads the industry for fast and precise measurements, even after repeated autoclaving or sterilization cycles at 140°C
  • Utilizes a silver-ion trap, keeping the reference junction clear even in the presence of sulfide-bearing solutions
  • Fully autoclavable or sterilizable in-situ
  • Gel electrolyte reduces maintenance
  • Resistant to poisoning substances
  • EHEDG certified
  • Pressure resistant up to 6 barg
  • Watertight connector IP 68
  • Integral temperature sensor permits automatic temperature compensation

Spezifikation

Spezifikation

VP cable connection; Pg 13.5 thread process connection
12 mm shaft
Bioprocesses, CIP, SIP
High alkali glass membrane; ceramic junction
Reference system of Argenthal with silver-ion trap
Operating pressure of 0 to 6 barg at 140°C
Analog pH/Temperature Electrode
425 mm
0 to 14
0°C to 140°C
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote