missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Influenza A H1N1 (A/California/04/2009) Neuraminidase His-tag Recombinant Protein

Artikelnummer. 17710779
Ansicht ändern
Click to view available options
Menge:
5 μg
Packungsgröße:
5 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17710779 5 μg 5 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17710779 Lieferant Invitrogen™ Lieferanten-Nr. A42583

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expiration Date: Expires one year from date of receipt when stored as instructed.

Influenza A virus is a major public health threat, killing more than 30,000 people per year in the USA. In early 2009, a novel swine-origin influenza A (H1N1) virus was identified in specimens obtained from patients in Mexico and the United States. The virus spread quickly around the world and on June 11, 2009, the World Health Organization declared it a pandemic. Influenza A virus has one of sixteen possible Hemagglutinin (HA) surface proteins and one of nine possible Neuraminidase (NA) surface proteins. The Hemagglutinin protein facilitates viral attachment while Neuraminidase is involved in viral release. These proteins also elicit immune responses that prevent infection or independently reduce viral replication. The genetic make-up of this swine flu virus is unlike any other: it is an H1N1 strain that combines a triple assortment first identified in 1998 including human, swine, and avian influenza with two new pig H3N2 virus genes from Eurasia, themselves of recent human origin. The distinct antigenic properties of the new swine influenza virus compared with seasonal influenza A (H1N1) virus suggest that human immunity against new swine influenza virus is limited, although the age distribution of reported cases suggests some degree of protection in older age groups.
TRUSTED_SUSTAINABILITY

Spezifikation

Zur Verwendung mit (Anwendung) Control, Bioactivity, Immunohistochemistry, Western Blot
Format Lyophilized
Zusammensetzung 20mM tris with 10% glycerol, 5% mannitol, 150mM NaCl, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.5
Molekulargewicht 53.6 kDa
Name Influenza A H1N1 (A/California/04/2009) Neuraminidase His-tag
pH-Bereich 7.5
Reinigungsverfahren Purified
Menge 5 μg
Lagerungsbedingungen -20°C or -80°C if preferred
Kennzeichnung RUO
Endotoxin-Konzentration <1.0 EU/μg
Gen-Alias H1N1 NA; Influenza H1N1
Gebräuchliche Bezeichnung Influenza A H1N1 NA
Biologische Aktivität Measured by its ability to cleave a fluorogenic substrate, 2'-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid. The specific activity is > 8, 000 pmoles/min/μg.
Produkttyp Protein
Konjugat Unconjugated
Spezies Influenza
Rekombinant Recombinant
Proteinmarkierung His-tag
Sequenz Influenza A virus H1N1 (A/California/04/2009) neuamindase, amino acids His36-Lys469 (Accession # ACP41107.1) with an N-terminal vasodilator-stimulated phosphoprotein tetramizeration domain and a C-terminal His-tag
Inhalt und Lagerung -20°Cor to +80°C if preferred
Aktivität Measured by its ability to cleave a fluorogenic substrate, 2'-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid. The specific activity is > 8, 000 pmoles/min/μg.
Endotoxin-Gehalt < 1 EU/μg (LAL)
Expressionssystem Insect cells
Proteinfamilie Other Proteins
Form Lyophilized
Forschungskategorie Virology
Reinheits- oder Qualitätsgrad >90% by SDS-PAGE
Protein Neuraminidase
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.