missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INF2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20466-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene represents a member of the formin family of proteins. It is considered a diaphanous formin due to the presence of a diaphanous inhibitory domain located at the N-terminus of the encoded protein. Studies of a similar mouse protein indicate that the protein encoded by this locus may function in polymerization and depolymerization of actin filaments. Mutations at this locus have been associated with focal segmental glomerulosclerosis 5.Spezifikation
INF2 | |
Polyclonal | |
Unconjugated | |
INF2 | |
C14orf151, C14orf173, HBEBP2 binding protein C, INF2, Inverted formin 2, pp9484 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64423, 70435 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q0GNC1, Q27J81 | |
INF2 | |
INF2 Fusion Protein Ag14302 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |