Learn More
Cytiva illustra™ ExoProStar™ S PCR- und Sequence Reaction Clean-up-Kit
Enzymatische PCR-Reinigungstechnologie mit Exonuclease I und Shrimp alkalischer Phosphatase zum Entfernen von nicht aufgenommenen Primern und dNTPs.
Marke: Cytiva US79500
Additional Details : Gewicht : 0.10000kg
Beschreibung
- Easy to automate
- Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
- Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
- Scalable for different reaction sizes
- No loss of PCR product
- Complete heat inactivation of the enzymes within 10 minutes
- Fast 15 minute protocol
- Updates existing enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs
- Improves digestion efficiency with no degradation of the target PCR product
- Sample is completely intact and ready for immediate use in manual or automated processes
Spezifikation
Enzymatic PCR clean up technology with Exonuclease I and Shrimp Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. | |
PCR and Sequence Reaction;Removes unincorporated primers and dNTPs; Improves digestion efficiency; No loss of PCR product | |
5000 | |
illustra ExoProStar S |