Learn More
IL-12 (p40/p70) Rat anti-Mouse, PE, Clone: C15.6, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 562038
Additional Details : Gewicht : 0.01000kg
Beschreibung
The C15.6 monoclonal antibody specifically binds to both free and complexed (homodimer p80 and heterodimer p70) forms of the p40 subunit of mouse interleukin-12 (IL-12). The immunogen used to generate the C15.6 hybridoma was recombinant mouse IL-12 p70 protein. p40 has also been described as a subunit of IL-23 and thus it is possible that the C15.6 antibody will crossreact with IL-23.
Spezifikation
IL-12 (p40/p70) | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
CHO-expressed recombinant mouse IL-12 p70 protein | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
C15.6 | |
PE | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 |
For Research Use Only.