Learn More
Invitrogen™ IL-10 Monoclonal Antibody (JES5-16E3), Functional Grade, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
2525.00 CHF - 16350.00 CHF
Spezifikation
| Antigen | IL-10 |
|---|---|
| Klon | JES5-16E3 |
| Konzentration | 1 mg/mL |
| Klassifikation | Monoclonal |
| Konjugat | Functional Grade |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|
15577806
|
Invitrogen™
16-7101-38 |
5 mg |
2525.00 CHF
5 Milligramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
17191472
|
Affymetrix eBioscience
16-7101-025 |
25 mg | |||||||
|
17101482
|
Affymetrix eBioscience
16-7101-050 |
50 mg | |||||||
Beschreibung
The JES5-16E3 antibody reacts with mouse interleukin-10 (IL-10). Mouse IL-10 is an ∽18 kDa factor also known as Cytokine Synthesis Inhibitory Factor (CSIF). In the mouse, Th2 cells, B1 cells, macrophages, and keratinocytes are the major cell subsets that produce IL-10. IL-10 inhibits synthesis of Th1 cytokines and proliferation of T cells, and acts as a costimulatory signal for mast cells, developing thymocytes and the Th2 response.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Spezifikation
| IL-10 | |
| 1 mg/mL | |
| Functional Grade | |
| Rat | |
| Mouse | |
| P18893 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| JES5-16E3 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| 16153 | |
| Primary | |
| 4°C | |
| IL10 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.