Learn More
IL-1 beta Monoclonal Antibody (CRM56), Functional Grade, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
148.00 CHF - 397.00 CHF
Spezifikation
Antigen | IL-1 beta |
---|---|
Klon | CRM56 |
Konzentration | 1 mg/mL |
Anwendungen | ELISA, Functional Assay, Neutralization |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15268737
|
Affymetrix eBioscience
16-7018-81 |
50 μg |
148.00 CHF
50 Mikrogramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15278737
|
Affymetrix eBioscience
16-7018-85 |
500 μg |
397.00 CHF
500 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Description: The CRM56 antibody reacts with human and baboon interleukin-1 beta; (IL-1 beta). IL-1 beta is a 17 kDa factor secreted primarily by monocytes. IL-1 has effects on T-helper cells, inducing IL-2 secretion and expression of IL-2 receptors. IL-1 has effects on B cells, promoting cell proliferation and immunoglobulin synthesis. Applications Reported: The CRM56 antibody has been reported for use in cytokine neutralization, and ELISA. Fluorochrome conjugated CRM56 is recommended for use in intracellular staining for flow cytometry. Functional Grade purified CRM56 antibody, cat. 16-7018, is recommended for use in functional assays. Applications Tested: The CRM56 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of human interleukin-1β (IL-1β) in combination with the biotin CRM57 (13-7016) antibody for detection and recombit human IL-1β (14-8018) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1 - 4 μg/mL. A standard curve consisting of doubling dilutions of the recombit standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. The Functional Grade Purified CRM56 antibody has been tested by LAL assay to verify low endotoxin and by bioassay to verify neutralization of IL-1β bioactivity. For in vitro neutralization, the CRM56 antibody at 2.
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.Spezifikation
IL-1 beta | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.2 | |
catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta | |
IL1B | |
Primary | |
4° C | |
IL1B |
CRM56 | |
ELISA, Functional Assay, Neutralization | |
Functional Grade | |
Mouse | |
Human | |
P01584 | |
3553 | |
IgG1 | |
Affinity chromatography | |
Antibody |
For Research Use Only.