missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IKBKB (IKK Beta), GST Tag Recombinant Protein
Human IKBKB (IKK Beta), GST Tag Recombinant Protein
Marke: Invitrogen PV3836
Additional Details : Gewicht : 0.00900kg
Beschreibung
Phosphorylation of serine residues on the I-kappa-B proteins by IKK kinases, including IKBKB, marks them for destruction via the ubiquitination pathway, allowing activation of the NF-kappa-B complex.Spezifikation
NP_001547 | |
3551, 4088 | |
10μg | |
IKK Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: Other | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate | |
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase beta | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
114 kDa | |
IKBKB (IKK Beta) | |
Human | |
GST Tag | |
Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic | |
Dry Ice | |
IKBKB | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
For Research Use Only. Not for use in diagnostic procedures.