missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Monoclonal Antibody (II/41), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marke: Affymetrix eBioscience 64-5790-80
Weitere Details : Gewicht : 0.01000kg
Beschreibung
Rat Monoclonal Antibody
IgM Monoclonal antibody specifically detects IgM in Mouse samples. It is validated for Flow CytometrySpezifikation
IgM | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P01872 | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
II/41 | |
Super Bright 645 | |
IGHM | |
IGHM | |
Affinity chromatography | |
RUO | |
16019 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |