missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgM Monoclonal Antibody (II/41), Super Bright™ 645, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
165.00 CHF - 389.00 CHF
Spezifikation
Antigen | IgM |
---|---|
Klon | II/41 |
Konzentration | 0.2 mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15993654
|
Affymetrix eBioscience
64-5790-82 |
100 μg |
389.00 CHF
100 Mikrogramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15983654
|
Affymetrix eBioscience
64-5790-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Rat Monoclonal Antibody
Mouse IgM is an immunoglobulin M antibody that is produced by B cells in response to an antigen. It is the first antibody produced during an immune response and is the largest immunoglobulin in the serum. Mouse IgM is composed of five heavy chains and five light chains, and it is involved in complement activation and agglutination of antigens. It plays a crucial role in the innate immune response and is important for neutralizing pathogens before the adaptive immune response is fully activated. Mouse IgM is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry.Spezifikation
IgM | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
16019 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
II/41 | |
Flow Cytometry | |
Super Bright 645 | |
Rat | |
Mouse | |
P01872 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |