Learn More
IgG3, κ, Clone: J606, Isotype Control, BD
Marke: BD Biosciences 562482
Additional Details : Gewicht : 0.00840kg
Beschreibung
Mouse IgG3 κ isotype control antibody is specific for the β-2 - 6-linked Fructosan. The transplantable plasmacytoma J606 was induced by intraperitoneal injection of mineral oils into BALB/c mice. This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488nm), green (532nm) and yellow-green (561nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20nm filter).
Host Species: Mouse
Clone: J606
Isotype: IgG3, κ
Flow Cytometry, Isotype Control
Spezifikation
J606 | |
PE-CF594 | |
Affinity Purified | |
4°C. Store in dark. Do not freeze. |
0.2mg/ml | |
Mouse | |
100 μg | |
IgG3 κ |