missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IgG1 Isotype Control, Alexa Fluor™ 647, Invitrogen™
Mouse Isotype Control
Marke: Invitrogen™ MG121
Additional Details : Gewicht : 0.50000kg
Beschreibung
Mouse IgG1 Isotype Control for Flow
Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Flow Cytometry.Spezifikation
Mouse IgG1 | |
Alexa Fluor 647 | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
purified | |
ASR | |
Isotype Control (Primary) | |
2-8°C | |
IgG1 | |
Wet Ice |
Flow Cytometry | |
PBS with 0.1% sodium azide | |
Mouse | |
500 μL | |
Whole Antibody | |
Not Applicable | |
Liquid | |
50 Tests | |
Alexa Fluor™ |