Learn More
IgG Heavy Chain Mouse anti-Rabbit, CoraLite 488, Clone: 1C9A5, Proteintech
Alle ansehen Proteintech ProdukteBeschreibung
The Heavy chain of Rabbit IgG antibody from Proteintech is a mouse monoclonal antibody to a protein of rabbit Heavy chain of Rabbit IgG. This antibody recognizes rabbit antigen. The Heavy chain of Rabbit IgG antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spezifikation
Spezifikation
| Antigen | IgG Heavy Chain |
| Anwendungen | Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | 1C9A5 |
| Konzentration | 2000 μg/mL |
| Konjugat | CoraLite 488 |
| Wirtsspezies | Mouse |
| Menge | 100 μL |
| Primär oder sekundär | Primary |
| Zielspezies | Rabbit |
| Mehr anzeigen |