missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Donkey anti-Human, Biotin, Secondary Antibody, Proteintech

Artikelnummer. 17690337 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
1 mL
200 μL
Unit Size:
1 Milliliter
200 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
17690337 200 μL 200 Mikroliter
17680337 1 mL 1 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17690337 Supplier Proteintech Supplier No. SA0000410200UL

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Donkey anti-Human Affinipure Secondary Antibody

Specifications

Antigen IgG(H+L)
Anwendungen ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry
Konjugat Biotin
Zusammensetzung 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Wirtsspezies Donkey
Reinigungsverfahren The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Menge 200 μL
Primär oder sekundär Secondary
Zielspezies Human
Inhalt und Lagerung Store at -20°C. Stable for one year after shipment.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.