Learn More
Syrian Hamster IgG Isotype Control, Biotin, eBioscience™, Invitrogen™
Syrian Hamster Isotype Control
Marke: Affymetrix eBioscience 13-4914-85
Additional Details : Gewicht : 0.25000kg
Beschreibung
Description: This hamster IgG fraction was purified by affinity chromatography from normal Golden Syrian hamster serum. Applications Reported: Golden Syrian hamster IgG isotype control has been reported for use in surface and intracellular flow cytometric analysis, and immunohistochemical staining. Applications Tested: This Golden Syrian hamster IgG isotype control has been tested by flow cytometric analysis of mouse splenocyte suspension. Isotype control should be used at the same concentration as experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.
The isotype of a primary antibody and the application it is being used in can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.Spezifikation
Syrian Hamster IgG | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Syrian Hamster | |
500 μg | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! | |
IgG |
Control, Flow Cytometry, Immunohistochemistry | |
Biotin | |
IgG; Immunoglobulin G; ImmunoglobulinG | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
For Research Use Only.