Learn More
IgG Fraction Mouse anti-Rabbit, HRP, Proteintech
Artikelnummer. 17204906 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
200 μL
Packungsgröße:
200 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17204906 200 μL 200 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17204906 Lieferant Proteintech Lieferanten-Nr. SA000017L200UL
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Monoclonal Antibody
IgG Fraction Monoclonal antibody specifically detects IgG Fraction in Rabbit samples. It is validated for ELISA, Western Blot.
Spezifikation
Antigen IgG Fraction
Anwendungen ELISA, Western Blot
Klassifikation Monoclonal
Konzentration 0.2 mg/mL
Konjugat HRP
Wirtsspezies Mouse
Menge 200 μL
Primär oder sekundär Secondary
Zielspezies Rabbit
Form Liquid
Isotype IgG1