Learn More
F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, eFluor™ 660, eBioscience™, Invitrogen™
F(ab')2-Goat Polyclonal Secondary Antibody
96.00 CHF - 227.00 CHF
Spezifikation
Antigen | Mouse IgG (H+L) |
---|---|
Konzentration | 0.2 mg/mL |
Klassifikation | Polyclonal |
Konjugat | eFluor 660 |
Form | Liquid |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15341900
|
Affymetrix eBioscience
50-4010-82 |
100 μg |
227.00 CHF
100 Mikrogramm |
Verfügbar ab: 29-04-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15331900
|
Affymetrix eBioscience
50-4010-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The goat anti-mouse IgG polyclonal antibody reacts with the heavy and light chains of mouse IgG, with minimal cross reactivity to human, rat, bovine, goat, hamster, and rabbit serum proteins. Reactivity has been shown to IgG1, IgG2a, IgG2b, and IgG3, as well as some reactivity to IgM. The antibody is a F(ab')2 fragment which contains the 2 Fab domains still linked through the hinge region of the immunoglobulin. Removal of the Fc region helps to reduce issues typically found with Fc-mediated binding. This polyclonal has been validated as a secondary reagent (its ability to recognize cells stained with a mouse IgG followed by this polyclonal antibody). Applications Reported: This polyclonal antibody has been reported for use in flow cytometric analysis, immunohistochemistry and immunocytochemistry. Applications Tested: This polyclonal antibody has been tested by flow cytometric analysis of normal human peripheral blood cells stained first with a mouse anti-human antibody. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. This polyclonal antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded sections that have been stained first with a mouse anti-human antibody.
Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
Mouse IgG (H+L) | |
Polyclonal | |
Liquid | |
RUO | |
Ig | |
Affinity chromatography |
0.2 mg/mL | |
eFluor 660 | |
Goat | |
Mouse | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.