missing translation for 'onlineSavingsMsg'
Learn More

IgD Monoclonal Antibody (IA6-2), Super Bright™ 600, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Affymetrix eBioscience 63-9868-42

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15973574

  • 399.00 CHF / 100 Stück
Verfügbar ab: 05-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Description: The G14D9 monoclonal antibody reacts with human Glycoprotein A Repetitions Predomit (GARP, also known as LRRC32 or Garpin). GARP is an approximately 80 kDa glycoprotein that is expressed on the cell surface. Using northern blot, RT-PCR or microarray analyses, the expression of GARP has been reported in placenta, lung, kidney, heart, ovary, liver, skeletal muscle, and pancreas. Protein expression has been observed on megakaryocytes, platelets and activated regulatory T (Treg) cells. The expression of GARP on the surface of activated Treg cells has been reported to be necessary for their suppressive function, possibly related to its role as a cell surface receptor for LAP/TGF beta. Applications Reported: This G14D9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This G14D9 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome.

This type I transmembrane protein is co-expressed on the surface of mature naive B cells with membrane IgM. IgD also exists as a soluble form in blood serum. Studies have demonstrated that IgD associates with the B cell receptor and participates in mediating signal transduction upon antigen binding.
Spezifikation

Spezifikation

IgD
Monoclonal
5 μL/Test
PBS with BSA and 0.09% sodium azide; pH 7.2
P01880
IGHD
Affinity chromatography
RUO
3495
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
IA6-2
Super Bright 600
IGHD
IGHD; immunoglobulin heavy constant delta
Mouse
100 Tests
Primary
Human
Antibody
IgG2a κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote