Learn More
IgA Rabbit anti-Mouse, Secondary Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen SA510308
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Mouse IgA Polyclonal antibody specifically detects Mouse IgA in Mouse samples. It is validated for ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
Spezifikation
| Mouse IgA | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Mouse | |
| Liquid |
| ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| Mouse IgA | |
| 1 mg | |
| Secondary | |
| 2-8°C | |
| IgG |