missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT122 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19304-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This cytoplasmic protein contains seven WD repeats and an AF-2 domain which function by recruiting coregulatory molecules and in transcriptional activation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Spezifikation
IFT122 | |
Polyclonal | |
Unconjugated | |
IFT122 | |
IFT122, SPG, WDR10, WDR10p, WDR140 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55764, 81896 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NWV3, Q9HBG6 | |
IFT122 | |
IFT122 Fusion Protein Ag6860 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |