Learn More
Invitrogen™ IFNAR1 Monoclonal Antibody (85228), PE
GREENER_CHOICE
Mouse Monoclonal Antibody
Marke:  Invitrogen™ MA523630
Artikelnummer. 15533322
  • 610.00 CHF / 100 Tests
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
In direct ELISAs, does not cross-react with recombinant human (rh) IFN- gamma R1, rhIFN-gamma R2, recombinant mouse IFN-alpha / beta R2, rhIL-10 R beta, rhIL-10 R alpha, or rhIL-20 R beta.
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system (Biron,2001, Kirkwood, 2002). These cytokines produce their effects by binding to the Type 1 Interferon-& Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1 (Kumar et al., 2003).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
IFNAR1
Monoclonal
PE
Ifnar1
alpha-type antiviral protein; AVP; beta-type antiviral protein; CD118; CRF2-1; cytokine receptor class-II member 1; Cytokine receptor family 2 member 1; H3K1; Ifar; IFN R1; IFNalpha/beta R1; IFN-alpha/beta receptor 1; IFN-alpha/betaR; IFNalpha/betaR1; IFN-alpha/betaR1; IFN-alpha/betaRalpha; IFNalpha/beta-Ralpha; IFN-alpha-REC; IFNAR; Ifnar1; IFNBR; IFNR1; IFN-R-1; Ifrc; INF-a receptor; Infar; interferon (alpha and beta) receptor 1; interferon (alpha, beta and omega) receptor 1; interferon alpha and beta receptor subunit 1; interferon alpha/beta receptor 1; interferon receptor 1; interferon-alpha/beta receptor 1; interferon-alpha/beta receptor alpha chain; interferon-beta receptor 1; type I interferon receptor 1
Mouse
Protein A/G
RUO
3454
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
85228
Buffered saline solution with ≤ 0.5 BSA and 0.09% sodium azide
P17181
Ifnar1
Sf 21-derived recombinant human IFN-alpha/beta R1 Lys28-Lys436.
100 Tests
Primary
Human
Antibody
IgG1
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen