Få mere at vide
Invitrogen™ Platin-ELISA-Kit für humanes IFN-gamma
Beskrivelse
Indeholder
- Aluminiumbeutel mit einer Mikrotiterplatte, beschichtet mit monoklonalem Antikörper gegen humanes IFN-gamma
- Biotin-Konjugat (monoklonaler Anti-Human-IFN-gamma-Antikörper)
- Streptavidin-HRP
- Human-IFN-gamma-Standard, gefriergetrocknet, 200 ng/ml nach Rekonstitution
- Probenverdünner
- Assaypuffer-Konzentrat 20x (PBS mit 1 % Tween 20 und 10 % BSA)
- Waschpuffer-Konzentrat 20x (PBS mit 1 % Tween 20)
- Substratlösung (Tetramethylbenzidin)
- Stopplösung (1M Phosphorsäure)
- Blauer Farbstoff
- Grüner Farbstoff
- Roter Farbstoff
- Klebefolien
The Human Interferon gamma (Hu IFNγ) ELISA quantitates Hu IFNγ in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IFNγ. Principle of the method The Human IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Tekniske data
Tekniske data
| Zugriffsnummer | Gehäuse P01579 |
| Assay-Bereich | 1.6 bis 100 pg/ml |
| Assay-Empfindlichkeit: | 0,99 pg/mL |
| Konjugat | Biotin |
| Produkttyp | ELISA-Kit |
| Probentyp | Plasma, Serum, Überstand |
| Zielspezies | Human |
| Zur Verwendung mit (Anwendung) | ELISA |
| Zur Verwendung mit (Geräte) | Kolorimetrisches Mikrotiterplatten-Lesegerät |
| Gen-ID (Entrez) | 3458 |
| Vis mere |
Nur für Forschungszwecke
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.