missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFI30 Polyclonal Antibody

Beschreibung
Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is a lysosomal thiol reductase that at low pH can reduce protein disulfide bonds. The enzyme is expressed constitutively in antigen-presenting cells and induced by gamma-interferon in other cell types. This enzyme has an important role in MHC class II-restricted antigen processing.
Spezifikation
Spezifikation
| Antigen | IFI30 |
| Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 0.2 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | PBS with trehalose and no preservative |
| Gen | IFI30 |
| Gen-Zugriffsnummer | P13284 |
| Gen-Alias | Gamma-interferon-inducible lysosomal thiol reductase; gamma-interferon-inducible protein IP-30; GILT; IFI30; IFI-30; IFI30, lysosomal thiol reductase; interferon gamma inducible protein 30; interferon gamma-inducible protein 30 preproprotein; interferon, gamma-inducible protein 30; IP30; IP-30; Legumaturain |
| Gensymbole | IFI30 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?