Learn More
Abnova™ IDI1 (Human) Recombinant Protein
Human IDI1 full-length ORF ( AAH05247.1, 1 a.a. - 228 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00003422-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MMPEINTNHLDKQQVQLLAEMCILIDENDNKIGAETKKNCHLNENIEKGLLHRAFSVFLFNTENKLLLQQRSDAKITFPGCFTNTCCSHPLSNPAELEESDALGVRRAAQRRLKAELGIPLEEVPPEEINYLTRIHYKAQSDGIWGEHEIDYILLVRKNVTLNPDPNEIKSYCYVSKEELKELLKKAASGEIKITPWFKIIAATFLFKWWDNLNHLNQFVDHEKIYRM
Spezifikation
AAH05247.1 | |
isopentenyl-diphosphate delta isomerase 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
IDI1 | |
GST |
3422 | |
Wheat germ expression system | |
25 μg | |
IPP1, IPPI1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |