missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | IDH1 |
---|---|
Konzentration | 0.17 mg/mL |
Anwendungen | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839993
|
Proteintech
12332-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829993
|
Proteintech
12332-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This nuclear enzyme is involved in processes such as chromosome condensation, chromatid separation, and the relief of torsional stress that occurs during DNA transcription and replication. It catalyzes the transient breaking and rejoining of two strands of duplex DNA which allows the strands to pass through one another, thus altering the topology of DNA. Two forms of this enzyme exist as likely products of a gene duplication event. The gene encoding this form, beta, is localized to chromosome 3 and the alpha form is localized to chromosome 17. The gene encoding this enzyme functions as the target for several anticancer agents and a variety of mutations in this gene have been associated with the development of drug resistance. Reduced activity of this enzyme may also play a role in ataxia-telangiectasia. Alternative splicing of this gene results in two transcript variants; however, the second variant has not yet been fully described.Spezifikation
IDH1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O75874, O88844, P41562 | |
15926, 24479, 3417 | |
IDH1 Fusion Protein Ag2986 | |
Primary | |
-20°C | |
IDH1 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IDCD, IDH, IDH1, IDP, IDPC, NADP(+) specific ICDH, Oxalosuccinate decarboxylase, PICD | |
IDH1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |