missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18475-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
ID1 is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. It has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.The protein encoded by this gene is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. The encoded protein has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.Spezifikation
ID1 | |
Polyclonal | |
Unconjugated | |
ID1 | |
bHLHb24, ID, ID1, Inhibitor of DNA binding 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15901, 25261, 3397 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20067, P41134, P41135 | |
ID1 | |
ID1 Fusion Protein Ag13359 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |