missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS (CD278) Armenian Hamster, PE-CF594, Clone: C398.4A, BD Horizon™
Armenian Hamster Monoclonal Antibody
730.00 CHF
Spezifikation
Antigen | ICOS (CD278) |
---|---|
Klon | C398.4A |
Konzentration | 0.2mg/mL |
Anwendungen | Flow Cytometry |
Klassifikation | Monoclonal |
Beschreibung
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.
ICOS (CD278) Monoclonal antibody specifically detects ICOS (CD278) in Human, Mouse, Rat , Rhesus Ma samples. It is validated for Flow cytometry.Spezifikation
ICOS (CD278) | |
0.2mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat, Rhesus Monkey | |
ICOS; AILIM; H4; Inducible T-cell costimulator; CVID1; Ly115 | |
IgG | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed. |
C398.4A | |
Flow Cytometry | |
PE-CF594 | |
Armenian Hamster | |
Cell Biology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse D10.G4.1 T-cell clone | |
Primary |
For Research Use Only.