Beschreibung
- ANSI A5/EN ISO E cut protection: 4x greater cut resistance
- Ultralight formulation: All-day comfort and dexterity ensured
- Touchscreen-compatible design: Improved productivity and safety
Spezifikation
Spezifikation
| Zertifizierung/Konformität | ANSI ABR 4, ANSI Cut A5, Dermatologically Tested, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020 |
| Beschichteter Bereich | Palm |
| Bauweise | Knit |
| Stulpenmaterial | HPPE, Tungsten, Spandex, Polyester |
| Beschreibung | Heightened defenses: Designed with INTERCEPT™ Cut Resistance Technology, and achieving ANSI A5/EN ISO E standards for cut resistance, HyFlex™ 11-755 industrial gloves guard against lacerations and prevent common workplace injuries. |
| Coating Material | Polyurethane |
| Farbe | Gray |
| Cut Resistance Rating | ANSI A5 |
| Hand | Specific |
| Latex Free | Yes |
| Mehr anzeigen |