missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-571 Nitrile Industrial Gloves, Palm-Coated

Artikelnummer. 17851019
Click to view available options
Größe:
5
6
7
8
9
10
11
12
Unit Size:
12 Paare
This item is not returnable. View return policy

Product Code. 17851019

Brand: Ansell 11571050

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The Hyflex™ 11-571 is the lightest ISO D/ ANSI A4-rated cut resistant glove offering 20% greater durability, providing enhanced comfort in abrasive conditions.

  • Ultra-lightweight design
  • Increased Durability for Extended Use: Improved FORTIX™ Technology delivers up to 20% more abrasion resistance for longer lasting handling in abrasive conditions (Compared to previous technology versions)
  • Heightened defenses: Developed with INTERCEPT™ Cut Resistance Technology using Next Generation HPPE single yarn, HyFlex™ 11-571 safety gloves achieve EN ISO D / ANSI A4 standards for cut resistance providing 2x more cut protection (Versus similar gloves made of standard HPPE yarn)
  • Touchscreen Compatible/ ESD: Allows the wearer to work multiple touchscreen devices while keeping gloves on
TRUSTED_SUSTAINABILITY

Specifications

Zertifizierung/Konformität ANSI ABR 4, ANSI Cut A4, Ecovadis label, EN ISO 21420:2020, EN 16350:2014, EN 388:2016 +A1:2018, Food Contact, OEKO-TEX™ Hohenstein, Dermatologically Tested
Beschichteter Bereich Palm
Bauweise Knit
Coating Material Nitrile
Farbe Gray
Cut Resistance Rating ANSI A4
Hand Specific
Latex Free Yes
Material HPPE, Carbon, Nylon, Spandex
Produkttyp Industrial Gloves
Größe 5
Norm EN 388 EN 388:2016 +A1:2018
Geschlecht Unisex
Art der Handschuhstulpe Knit
Auskleidungsmaterial HPPE, Carbon, Nylon, Spandex
Modell 11571
Yarn Gauge 15-gauge
Länge (metrisch) 205 to 272 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.