Learn More
Hydroxynorketamine Hydrochloride (100 μg/mL in Methanol), TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Hydroxynorketamine Hydrochloride |
| CAS | 2173991-73-4 |
| Summenformel | C12H15Cl2NO2 |
| Menge | 5 x 1 mL |
| Synonym | 2-Amino-2-(2-chlorophenyl)-6-hydroxy-cyclohexanone Hydrochloride |
| InChI-Formel | InChI=1S/C12H14ClNO2.ClH/c13-9-5-2-1-4-8(9)12(14)7-3-6-10(15)11(12)16;/h1-2,4-5,10,15H,3,6-7,14H2;1H |
| SMILES | NC(C1=O)(CCCC1O)C2=CC=CC=C2Cl.Cl |
| Molekulargewicht (g/mol) | 276.16 |
| Formelmasse | 275.048 |
| Concentration or Composition (by Analyte or Components) | Hydroxynorketamine Hydrochloride 100 μg/mL |