missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™-Wasser-in-Methanol 5.0 , Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Menge:
500 ml
1 l
6 x 1 L
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Spécification
Chemischer Name oder Material | HYDRANAL™-Water-in-methanol 5.0 |
Flammpunkt | 9°C (48.2°F) |
Menge | 6 x 1 L |
UN-Nummer | UN1230 |
Formelmasse | 32.04 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'