missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IL-1 alpha protein

Product Code. 17620087 Shop All Proteintech Products
missing translation for 'orderingAttributeHoverText'
Menge:
10 μg
100 μg
1000 μg
missing translation for 'unitSize'
10 Mikrogramm
100 Mikrogramm
1000 Mikrogramm
This item is not returnable. View return policy

Product Code. 17620087

missing translation for 'mfr': Proteintech HZ13201000UG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

HumanKine™ recombinant protein

Specifications

Zugriffsnummer P01583
Zusammensetzung 1 x PBS, See Certificate of Analysis for details
Gen-ID (Entrez) 3552
Molekulargewicht 22 kDa, monomer, glycosylated
Menge 100 μg
Quelle HEK293
Lagerungsbedingungen Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin-Konzentration <1 EU/μg
Spezifische Reaktivität Human|Mouse
Rekombinant Recombinant
Rekonstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Aktivität 0.075-0.375 ng/mL EC50
Form Lyophilized
Reinheits- oder Qualitätsgrad >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.