missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech™ HumanKine™ Recombinant Human IFN Alpha 2A Protein
Alle ansehen Proteintech Produkte
missing translation for 'orderingAttributeHoverText'
Menge:
10 μg
100 μg
1000 μg
missing translation for 'unitSize'
10 Mikrogramm
100 Mikrogramm
1000 Mikrogramm
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Specifications
| Gen-ID (Entrez) | 3440 |
| Molekulargewicht | 16 kDa, monomer, glycosylated |
| Menge | 100 μg |
| Spezies | Human |
| Rekonstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
| Inhalt und Lagerung | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
| Aktivität | Typically ≤ 0.4 ng/mL EC50 |
| Expressionssystem | Human embryonic kidney 293 (HEK293) human cell expression system |
| Form | Lyophilized |
| Reinheits- oder Qualitätsgrad | > 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction