missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNFX1 Partial ORF (NP_066363.1, 50 a.a. - 150 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00057169-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: PGRHPRANNHPAAYWQREERFRAMGRNPHQGRRNQEGHASDEARDQRHDQENDTRWRNGNQDCRNRRPPWSNDNFQQWRTPHQKPTEQPQQAKKLGYKFLESpezifikation
NP_066363.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.85kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PGRHPRANNHPAAYWQREERFRAMGRNPHQGRRNQEGHASDEARDQRHDQENDTRWRNGNQDCRNRRPPWSNDNFQQWRTPHQKPTEQPQQAKKLGYKFLE | |
RUO | |
ZNFX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57169 | |
ZNFX1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ39275/MGC131926 | |
ZNFX1 | |
Recombinant | |
wheat germ expression system |