missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF549 Full-length ORF (AAH60863.1, 1 a.a. - 640 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | AAH60863.1 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 256051 |
Molekulargewicht | 100.8kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16119703
|
Abnova™
H00256051-P01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 06-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16129703
|
Abnova™
H00256051-P01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 06-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Sequence: MAEAALVNTPQIPMVTEEFVKPSQGHVTFEDIAVYFSQEEWGLLDEAQRCLYHDVMLENFSLMASVGCLHGIEAEEAPSEQTLSAQGVSQARTPKLGPSIPNAHSCEMCILVMKDILYLSEHQGTLPWQKPYTSVASGKWFSFGSNLQQHQNQDSGEKHIRKEESSALLLNSCKIPLSDNLFPCKDVEKDFPTILGLLQHQTTHSRQEYAHRSRETFQQRRYKCEQVFNEKVHVTEHQRVHTGEKAYKRREYGKSLNSKYLFVEHQRTHNAEKPYVCNICGKSFLHKQTLVGHQQRIHTRERSYVCIECGKSLSSKYSLVEHQRTHNGEKPYVCNVCGKSFRHKQTFVGHQQRIHTGERPYVCMECGKSFIHSYDRIRHQRVHTGEGAYQCSECGKSFIYKQSLLDHHRIHTGERPYECKECGKAFIHKKRLLEHQRIHTGEKPYVCIICGKSFIRSSDYMRHQRIHTGERAYECSDCGKAFISKQTLLKHHKIHTRERPYECSECGKGFYLEVKLLQHQRIHTREQLCECNECGKVFSHQKRLLEHQKVHTGEKPCECSECGKCFRHRTSLIQHQKVHSGERPYNCTACEKAFIYKNKLVEHQRIHTGEKPYECGKCGKAFNKRYSLVRHQKVHITEEPSpezifikation
AAH60863.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
100.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAEAALVNTPQIPMVTEEFVKPSQGHVTFEDIAVYFSQEEWGLLDEAQRCLYHDVMLENFSLMASVGCLHGIEAEEAPSEQTLSAQGVSQARTPKLGPSIPNAHSCEMCILVMKDILYLSEHQGTLPWQKPYTSVASGKWFSFGSNLQQHQNQDSGEKHIRKEESSALLLNSCKIPLSDNLFPCKDVEKDFPTILGLLQHQTTHSRQEYAHRSRETFQQRRYKCEQVFNEKVHVTEHQRVHTGEKAYKRREYGKSLNSKYLFVEHQRTHNAEKPYVCNICGKSFLHKQTLVGHQQRIHTRERSYVCIECGKSLSSKYSLVEHQRTHNGEKPYVCNVCGKSFRHKQTFVGHQQRIHTGERPYVCMECGKSFIHSYDRIRHQRVHTGEGAYQCSECGKSFIYKQSLLDHHRIHTGERPYECKECGKAFIHKKRLLEHQRIHTGEKPYVCIICGKSFIRSSDYMRHQRIHTGERAYECSDCGKAFISKQTLLKHHKIHTRERPYECSECGKGFYLEVKLLQHQRIHTREQLCECNECGKVFSHQKRLLEHQKVHTGEKPCECSECGKCFRHRTSLIQHQKVHSGERPYNCTACEKAFIYKNKLVEHQRIHTGEKPYECGKCGKAFNKRYSLVRHQKVHITEEP | |
RUO | |
ZNF549 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
256051 | |
ZNF549 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ34917 | |
ZNF549 | |
Recombinant | |
wheat germ expression system |