missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF354A Partial ORF (NP_005640, 106 a.a. - 215 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00006940-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: GLILKRSNRNVPWDLKLEKPYIYEGRLEKKQDKKGSFQIVSATHKKIPTIERSHKNTELSQNFSPKSVLIRQQILPREKTPPKCEIQGNSLKQNSQLLNQPKITADKRYKSpezifikation
NP_005640 | |
Liquid | |
6940 | |
ZNF354A (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EZNF/HKL1/KID-1/KID1/TCF17 | |
ZNF354A | |
Yes | |
wheat germ expression system |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GLILKRSNRNVPWDLKLEKPYIYEGRLEKKQDKKGSFQIVSATHKKIPTIERSHKNTELSQNFSPKSVLIRQQILPREKTPPKCEIQGNSLKQNSQLLNQPKITADKRYK | |
RUO | |
ZNF354A | |
Wheat Germ (in vitro) | |
GST |