missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF274 Full-length ORF (NP_598009.1, 1 a.a. - 653 a.a.) Recombinant Protein with GST-tag at N-terminal
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Beschreibung
This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals. [provided by RefSeq]
Sequence: MASRLPTAWSCEPVTFEDVTLGFTPEEWGLLDLKQKSLYREVMLENYRNLVSVEHQLSKPDVVSQLEEAEDFWPVERGIPQDTIPEYPELQLDPKLDPLPAESPLMNIEVVEVLTLNQEVAGPRNAQIQALYAEDGSLSADAPSEQVQQQGKHPGDPEAARQRFRQFRYKDMTGPREALDQLRELCHQWLQPKARSKEQILELLVLEQFLGALPVKLRTWVESQHPENCQEVVALVEGVTWMSEEEVLPAGQPAEGTTCCLEVTAQQEEKQEDAAICPVTVLPEEPVTFQDVAVDFSREEWGLLGPTQRTEYRDVMLETFGHLVSVGWETTLENKELAPNSDIPEEEPAPSLKVQESSRDCALSSTLEDTLQGGVQEVQDTVLKQMESAQEKDLPQKKHFDNRESQANSGALDTNQVSLQKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHTGERPYVCQDCGKGFVQSSSLTQHQRVHSGERPFECQECGRTFNDRSAISQHLRTHTGAKPYKCQDCGKAFRQSSHLIRHQRTHTGERPYACNKCGKAFTQSSHLIGHQRTHNRTKRKKKQPTS
Spezifikation
Spezifikation
Zugriffsnummer | NP_598009.1 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 10782 |
Molekulargewicht | 100.6kDa |
Name | ZNF274 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Immunogen | MASRLPTAWSCEPVTFEDVTLGFTPEEWGLLDLKQKSLYREVMLENYRNLVSVEHQLSKPDVVSQLEEAEDFWPVERGIPQDTIPEYPELQLDPKLDPLPAESPLMNIEVVEVLTLNQEVAGPRNAQIQALYAEDGSLSADAPSEQVQQQGKHPGDPEAARQRFRQFRYKDMTGPREALDQLRELCHQWLQPKARSKEQILELLVLEQFLGALPVKLRTWVESQHPENCQEVVALVEGVTWMSEEEVLPAGQPAEGTTCCLEVTAQQEEKQEDAAICPVTVLPEEPVTFQDVAVDFSREEWGLLGPTQRTEYRDVMLETFGHLVSVGWETTLENKELAPNSDIPEEEPAPSLKVQESSRDCALSSTLEDTLQGGVQEVQDTVLKQMESAQEKDLPQKKHFDNRESQANSGALDTNQVSLQKIDNPESQANSGALDTNQVLLHKIPPRKRLRKRDSQVKSMKHNSRVKIHQKSCERQKAKEGNGCRKTFSRSTKQITFIRIHKGSQVCRCSECGKIFRNPRYFSVHKKIHTGERPYVCQDCGKGFVQSSSLTQHQRVHSGERPFECQECGRTFNDRSAISQHLRTHTGAKPYKCQDCGKAFRQSSHLIRHQRTHTGERPYACNKCGKAFTQSSHLIGHQRTHNRTKRKKKQPTS |
Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Abnova™ Human ZNF274 Full-length ORF (NP_598009.1, 1 a.a. - 653 a.a.) Recombinant Protein with GST-tag at N-terminal >
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur