missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF265 Partial ORF (NP_005446, 1 a.a. - 106 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00009406-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MSTKNFRVSDGDWICPDKKCGNVNFARRTSCNRCGREKTTEAKMMKAGGTEIGKTLAEKSRGLFSANDWQCKTCSNVNWARRSECNMCNTPKYAKLEERTGYGGGFSpezifikation
NP_005446 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.4kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTKNFRVSDGDWICPDKKCGNVNFARRTSCNRCGREKTTEAKMMKAGGTEIGKTLAEKSRGLFSANDWQCKTCSNVNWARRSECNMCNTPKYAKLEERTGYGGGF | |
RUO | |
ZRANB2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9406 | |
ZNF265 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J1831/DKFZp686N09117/FLJ41119/ZIS/ZIS1/ZIS2/ZNF265 | |
ZRANB2 | |
Recombinant | |
wheat germ expression system |