missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF253 Partial ORF (NP_066385.2, 91 a.a. - 180 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00056242-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: QNSFQIGMLRRYEECRHDNLQLKKGCKSVGEHKVHKGGYNGLNQCLTTTQKEIFQCDKYGKVFHKFSNSNTYKTRHTGINLFKCIICGKASpezifikation
NP_066385.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
QNSFQIGMLRRYEECRHDNLQLKKGCKSVGEHKVHKGGYNGLNQCLTTTQKEIFQCDKYGKVFHKFSNSNTYKTRHTGINLFKCIICGKA | |
RUO | |
ZNF253 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56242 | |
ZNF253 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMZF-1/BMZF1/FLJ90391/ZNF411 | |
ZNF253 | |
Recombinant | |
wheat germ expression system |