missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF101 Partial ORF (NP_149981.2, 112 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00094039-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: LRHSFLDRHMRAHAGHKRSECGGEWRETPRKQKQHGKASISPSSGARRTVTPTRKRPYECKVCGKAFNSPNLFQIHQRTHTGKRSYKCRESpezifikation
NP_149981.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LRHSFLDRHMRAHAGHKRSECGGEWRETPRKQKQHGKASISPSSGARRTVTPTRKRPYECKVCGKAFNSPNLFQIHQRTHTGKRSYKCRE | |
RUO | |
ZNF101 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
94039 | |
ZNF101 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp570I0164/HZF12/MGC149565/MGC149566 | |
ZNF101 | |
Recombinant | |
wheat germ expression system |