Learn More
Abnova™ Human ZNF10 Partial ORF (NP_056209.2, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007556-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene contains a C2H2 zinc finger, and has been shown to function as a transcriptional repressor. The Kruppel-associated box (KRAB) domain of this protein is found to be responsible for its transcriptional repression activity. RING finger containing protein TIF1 was reported to interact with the KRAB domain, and may serve as a mediator for the repression activity of this protein. [provided by RefSeq]
Sequence: MDAKSLTAWSRTLVTFKDVFVDFTREEWKLLDTAQQIVYRNVMLENYKNLVSLGYQLTKPDVILRLEKGEEPWLVEREIHQETHPDSETAFEIKSSVSSpezifikation
NP_056209.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDAKSLTAWSRTLVTFKDVFVDFTREEWKLLDTAQQIVYRNVMLENYKNLVSLGYQLTKPDVILRLEKGEEPWLVEREIHQETHPDSETAFEIKSSVS | |
RUO | |
ZNF10 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7556 | |
ZNF10 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KOX1 | |
ZNF10 | |
Recombinant | |
wheat germ expression system |