missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZIC3 Full-length ORF (NP_003404.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00007547-P01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16131942

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 07-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs. [provided by RefSeq]

Sequence: MTMLLDGGPQFPGLGVGSFGAPRHHEMPNREPAGMGLNPFGDSTHAAAAAAAAAAFKLSPAAAHDLSSGQSSAFTPQGSGYANALGHHHHHHHHHHHTSQVPSYGGAASAAFNSTREFLFRQRSSGLSEAASGGGQHGLFAGSASSLHAPAGIPEPPSYLLFPGLHEQGAGHPSPTGHVDNNQVHLGLRGELFGRADPYRPVASPRTDPYAAGAQFPNYSPMNMNMGVNVAAHHGPGAFFRYMRQPIKQELSCKWIDEAQLSRPKKSCDRTFSTMHELVTHVTMEHVGGPEQNNHVCYWEECPREGKSFKAKYKLVNHIRVHTGEKPFPCPFPGCGKIFARSENLKIHKRTHTGEKPFKCEFEGCDRRFANSSDRKKHMHVHTSDKPYICKVCDKSYTHPSSLRKHMKVHESQGSDSSPAASSGYESSTPPAIASANSKDTTKTPSAVQTSTSHNPGLPPNFNEWYV
Spezifikation

Spezifikation

NP_003404.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
77kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HTX/HTX1/ZNF203
ZIC3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
7547
ZIC3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTMLLDGGPQFPGLGVGSFGAPRHHEMPNREPAGMGLNPFGDSTHAAAAAAAAAAFKLSPAAAHDLSSGQSSAFTPQGSGYANALGHHHHHHHHHHHTSQVPSYGGAASAAFNSTREFLFRQRSSGLSEAASGGGQHGLFAGSASSLHAPAGIPEPPSYLLFPGLHEQGAGHPSPTGHVDNNQVHLGLRGELFGRADPYRPVASPRTDPYAAGAQFPNYSPMNMNMGVNVAAHHGPGAFFRYMRQPIKQELSCKWIDEAQLSRPKKSCDRTFSTMHELVTHVTMEHVGGPEQNNHVCYWEECPREGKSFKAKYKLVNHIRVHTGEKPFPCPFPGCGKIFARSENLKIHKRTHTGEKPFKCEFEGCDRRFANSSDRKKHMHVHTSDKPYICKVCDKSYTHPSSLRKHMKVHESQGSDSSPAASSGYESSTPPAIASANSKDTTKTPSAVQTSTSHNPGLPPNFNEWYV
RUO
ZIC3
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote