Learn More
Abnova™ Human ZFYVE1 Full-length ORF (NP_067083.1, 1 a.a. - 777 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00053349-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
The FYVE domain mediates the recruitment of proteins involved in membrane trafficking and cell signaling to phosphatidylinositol 3-phosphate (PtdIns(3)P)-containing membranes. This gene encodes a protein which contains two zinc-binding FYVE domains in tandem. This protein displays a predominantly Golgi, endoplasmic reticulum and vesicular distribution. Alternatively spliced transcript variants have been found for this gene, and they encode two isoforms with different sizes. [provided by RefSeq]
Sequence: MSAQTSPAEKGLNPGLMCQESYACSGTDEAIFECDECCSLQCLRCEEELHRQERLRNHERIRLKPGHVPYCDLCKGLSGHLPGVRQRAIVRCQTCKINLCLECQKRTHSGGNKRRHPVTVYNVSNLQESLEAEEMDEETKRKKMTEKVVSFLLVDENEEIQVTNEEDFIRKLDCKPDQHLKVVSIFGNTGDGKSHTLNHTFFYGREVFKTSPTQESCTVGVWAAYDPVHKVAVIDTEGLLGATVNLSQRTRLLLKVLAISDLVIYRTHADRLHNDLFKFLGDASEAYLKHFTKELKATTARCGLDVPLSTLGPAVIIFHETVHTQLLGSDHPSEVPEKLIQDRFRKLGRFPEAFSSIHYKGTRTYNPPTDFSGLRRALEQLLENNTTRSPRHPGVIFKALKALSDRFSGEIPDDQMAHSSFFPDEYFTCSSLCLSCGVGCKKSMNHGKEGVPHEAKSRCRYSHQYDNRVYTCKACYERGEEVSVVPKTSASTDSPWMGLAKYAWSGYVIECPNCGVVYRSRQYWFGNQDPVDTVVRTEIVHVWPGTDGFLKDNNNAAQRLLDGMNFMAQSVSELSLGPTKAVTSWLTDQIAPAYWRPNSQILSCNKCATSFKDNDTKHHCRACGEGFCDSCSSKTRPVPERGWGPAPVRVCDNCYEARNVQLAVTEAQVDDEGGTLIARKVGEAVQNTLGAVVTAIDIPLGLVKDAARPAYWVPDHEILHCHNCRKEFSIKLSKHHCRACGQGFCDECSHDRRAVPSRGWDHPVRVCFNCNKKPGDLSpezifikation
NP_067083.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
113.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFCP1/KIAA1589/TAFF1/ZNFN2A1 | |
ZFYVE1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
53349 | |
ZFYVE1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSAQTSPAEKGLNPGLMCQESYACSGTDEAIFECDECCSLQCLRCEEELHRQERLRNHERIRLKPGHVPYCDLCKGLSGHLPGVRQRAIVRCQTCKINLCLECQKRTHSGGNKRRHPVTVYNVSNLQESLEAEEMDEETKRKKMTEKVVSFLLVDENEEIQVTNEEDFIRKLDCKPDQHLKVVSIFGNTGDGKSHTLNHTFFYGREVFKTSPTQESCTVGVWAAYDPVHKVAVIDTEGLLGATVNLSQRTRLLLKVLAISDLVIYRTHADRLHNDLFKFLGDASEAYLKHFTKELKATTARCGLDVPLSTLGPAVIIFHETVHTQLLGSDHPSEVPEKLIQDRFRKLGRFPEAFSSIHYKGTRTYNPPTDFSGLRRALEQLLENNTTRSPRHPGVIFKALKALSDRFSGEIPDDQMAHSSFFPDEYFTCSSLCLSCGVGCKKSMNHGKEGVPHEAKSRCRYSHQYDNRVYTCKACYERGEEVSVVPKTSASTDSPWMGLAKYAWSGYVIECPNCGVVYRSRQYWFGNQDPVDTVVRTEIVHVWPGTDGFLKDNNNAAQRLLDGMNFMAQSVSELSLGPTKAVTSWLTDQIAPAYWRPNSQILSCNKCATSFKDNDTKHHCRACGEGFCDSCSSKTRPVPERGWGPAPVRVCDNCYEARNVQLAVTEAQVDDEGGTLIARKVGEAVQNTLGAVVTAIDIPLGLVKDAARPAYWVPDHEILHCHNCRKEFSIKLSKHHCRACGQGFCDECSHDRRAVPSRGWDHPVRVCFNCNKKPGDL | |
RUO | |
ZFYVE1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |