missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZC3HAV1 Partial ORF (NP_078901.3, 601 a.a. - 699 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00056829-Q01.10ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16128816

  • 361.00 CHF / 10 Mikrogramm
Verfügbar ab: 07-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a CCCH-type zinc finger protein that is thought to prevent infection by retroviruses. Studies of the rat homolog indicate that the protein may primarily function to inhibit viral gene expression and induce an innate immunity to viral infection. Alternative splicing occurs at this locus and two variants, each encoding distinct isoforms, are described. [provided by RefSeq]

Sequence: SVFTTKWIWYWKNESGTWIQYGEEKDKRKNSNVDSSYLESLYQSCPRGVVPFQAGSRNYELSFQGMIQTNIASKTQKDVIRRPTFVPQWYVQQMKRGPE
Spezifikation

Spezifikation

NP_078901.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SVFTTKWIWYWKNESGTWIQYGEEKDKRKNSNVDSSYLESLYQSCPRGVVPFQAGSRNYELSFQGMIQTNIASKTQKDVIRRPTFVPQWYVQQMKRGPE
RUO
ZC3HAV1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
56829
ZC3HAV1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686F2052/DKFZp686H1869/DKFZp686O19171/FLB6421/FLJ13288/MGC48898/ZAP/ZC3H2/ZC3HDC2
ZC3HAV1
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote